Methylenblau (Neisser) für die Mikroskopie, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 3,7-Bis(dimethylamino)phenothiaziniumchlorid, N,N,N′,N′-Tetramethylthioninchlorid, Methylthioniniumchlorid

M-Clarity™ Qualitätsstufe = MQ100

01363-100ML-F
01363-100ML-FEA 43.3 EUR
01363-100ML-F
Methylenblau (Neisser) für die Mikroskopie, Sigma-Aldrich®
Methylenblau

Methylene blue, Neisser formulation. Well developed granules of volutin (polyphosphate) may be seen in unstained wet preparations as round refractive bodies within the bacterial cytoplasm. With methylene blue they stain metachromatically, a reddish purple color.


  • Hematology: suitable
  • Histology: suitable


Components
Methylene blue 1 g/L, ethanol (95 %) 50 mL/L, glacial acetic acid 50 mL/L in water

Formel: C₁₆H₁₈ClN₃S
Molecular Weight: 319,86 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150008
CAS-Nummer: 61-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR