VWR®, Glycylglycine (Gly-Gly)
Lieferant: VWR Chemicals
0137-1KGEA
892
EUR
0137-1KG
VWR®, Glycylglycine (Gly-Gly)
Proteine und Peptide
Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH.
- Loss on drying: ≤0,2%
- Solubility (10%, water): Pass
- EINECS: 209-127-8
- GHS symbol and signal word: GHS07 and warning
Glycylglycine (Gly-Gly) contains ≤0,02% ammonia, ≤0,0001% arsenic, ≤0,2% ash, ≤0,02% chloride, ≤0,002% heavy metals (as Pb), ≤0,001% iron and ≤0,03% sulphate.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...