Supelco® Organic Reference Standard, Acetone-2,4-Dnph
Lieferant: Merck
01378-50MG
01378-50MGEA 427 EUR
01378-50MG
Supelco® Organic Reference Standard, Acetone-2,4-Dnph
Standards Organische Standards
  • GHS signal: GHS07
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P305 + P351 + P338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bf7d391a94a25f2bbb80c3e33abd833043690a1&vid=7bf7d391a94a25f2bbb80c3e33abd833043690a1&prid=web&clid=SAPDAM