Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3) ≥98%, Ultrareine Qualität

Lieferant: VWR Chemicals

0145-250MGEA 123 EUR
0145-250MG
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3) ≥98%, Ultrareine Qualität
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3)

VWR offers riboneucleotides for amplification of RNA in in vitro transcription.

Formel: C₉H₁₂N₂Na₃O₁₅P₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00044310
CAS-Nummer: 19817-92-6
EINECS: 243-347-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR