Chinesische Tusch, grün, Q Path® Färbemittel für Histologie

Lieferant: VWR Q-Path Chemicals

01816201.EA 50.9 EUR
01816201.
Chinesische Tusch, grün, Q Path® Färbemittel für Histologie
Chinesische Tusch

Q Path Inks for Pathology are inks for marking and identifying histological samples without altering the tissues.


  • Very long lasting non removable ink
  • Product ready for use


Usage
1. Shake the bottle well before opening
2. The ink is applied using a paintbrush or a swab
3. Close the bottle properly after use


Composition: Water, pigment and glycerine.

Bestellen

Spezifikation Testergebnisse

Appearance Green liquid
Colour of solution Passes test
Density (20/4) 1.190 --> 1.370

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR