Chinesische Tusch, rot, Q Path® Färbemittel für Histologie
Lieferant: VWR Q-Path Chemicals
01816204.EA
85.2
EUR
01816204.
Chinesische Tusch, rot, Q Path® Färbemittel für Histologie
Chinesische Tusch
Q Path Inks for Pathology are inks for marking and identifying histological samples without altering the tissues.
- Very long lasting non removable ink
- Product ready for use
Usage
1. Shake the bottle well before opening
2. The ink is applied using a paintbrush or a swab
3. Close the bottle properly after use
Composition: Water, pigment and glycerine.
Spezifikation Testergebnisse
Appearance | Red liquid |
Colour of solution | Passes test |
Density (20/4) | 1.130 --> 1.170 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...