CYPMPO ≥97% für ESR-Spektroskopie, Supelco®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
01872-1MG
01872-1MGEA
161
EUR
01872-1MG
CYPMPO ≥97% für ESR-Spektroskopie, Supelco®
CYPMPO
CYPMPO is designed for radical detection. It is a free radical spin trap with excellent trapping capabilites toward hydroxyl and superoxide radicals in biological and chemical systems.
Formel:
C₁₀H₁₈NO₄P Molecular Weight: 247.23 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD11045266 CAS-Nummer: 934182-09-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...