Milchsäure 85%, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: DL-Milchsäure

0194-01EA 123.73 EUR
0194-01
Milchsäure 85%, BAKER ANALYZED® ACS, J.T.Baker®
Milchsäure

The reagent generally available is a mixture of lactic acid (CH₃CHOHCOOH) and lactic acid lactate (C₆H₁₀O₅).

Formel: H₃CCH(OH)COOH
Molecular Weight: 90,08 g/mol
Siedepunkt: 122 °C (20 hPa)
Dichte: 1,20 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00004520
CAS-Nummer: 50-21-5
EINECS: 200-018-0
UN: 3265
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 85.0 - 90.0%
For Laboratory,Research,or Manufacturing Use ≤0.02%
ACS - Assay (as CH₃CHOHCOOH) 85.0 - 90.0 %
ACS - Residue after Ignition ≤ 0.02 %
ACS - Chloride (Cl) ≤ 0.001 %
ACS - Sulfate (SO₄) ≤ 0.002 %
ACS - Substances Darkened by H₂SO₄ Passes Test
Trace Impurities - ACS - Heavy Metals (as Pb) ≤ 5 ppm
Trace Impurities - Iron (Fe) ≤ 5 ppm
The reagent generally available is a mixture of lactic acid (CH₃CHOHCOOH) andlactic acid lactate (C₆H₁₀O₅).

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR