Natriumhydroxid Monohydrat 99,9995 Metals basis, TraceSELECT™ Trace_analysis, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

01968-100G 01968-25G
01968-100GEA 239.38 EUR
01968-100G 01968-25G
Natriumhydroxid Monohydrat 99,9995 Metals basis, TraceSELECT™ Trace_analysis, Fluka™
Natriumhydroxid Monohydrat

TraceSELECT™ Ultra and TraceSELECT


  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formel: HNaO·1H₂O
Molecular Weight: 58.012 g/mol
Siedepunkt: 1.390 °C (1.013 hPa) (wasserfrei)
Schmelzpunkt: 319 °C (wasserfrei)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149229
CAS-Nummer: 12200-64-5
EINECS: 215-185-5
UN: 1823
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Barium (Ba) Max. 0.5 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cerium (Ce) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Europium (Eu) Max. 0.01 ppm
Iron (Fe) Max. 0.05 ppm
Mercury (Hg) Max. 1 ppb
Iridium (Ir) Max. 0.5 ppm
Potassium (K) Max. 2 ppm
Lanthanum (La) Max. 0.01 ppm
Lithium (Li) Max. 0.2 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Palladium (Pd) Max. 0.5 ppm
Platinum (Pt) Max. 0.5 ppm
Rhodium (Rh) Max. 0.5 ppm
Ruthenium (Ru) Max. 0.5 ppm
Antimony (Sb) Max. 0.05 ppm
Scandium (Sc) Max. 0.01 ppm
Selenium (Se) Max. 0.01 ppm
Samarium (Sm) Max. 0.01 ppm
Strontium (Sr) Max. 0.5 ppm
Thallium (Tl) Max. 0.01 ppm
Vanadium (V) Max. 0.1 ppm
Yttrium (Y) Max. 0.01 ppm
Ytterbium (Yb) Max. 0.01 ppm
Zinc (Zn) Max. 0.05 ppm
Silicate (as SiO₂) Max. 5 ppm
Bromide (Br) Max. 0.5 ppm
Chloride (Cl) Max. 1 ppm
Fluoride (F) Max. 0.5 ppm
Nitrite (NO₂) Max. 0.5 ppm
Nitrate (NO₃) Max. 1 ppm
Phosphate (PO₄) Max. 0.05 ppm
Sulfate (SO₄) Max. 2.5 ppm
Total N Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR