Kaliumhydrogensulfat, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Kaliumbisulfat

0198.0500EA 54.49 EUR
0198.0500
Kaliumhydrogensulfat, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Kaliumhydrogensulfat
Formel: KHSO₄
Schmelzpunkt: 214 °C
Dichte: 2,32 g/cm³ (20 °C)
MDL: MFCD00011404
CAS-Nummer: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 13,07695

Bestellen

Spezifikation Testergebnisse

Acidity (as H2SO4) 35.0 - 37.0 %
Calcium and Magnesium Precipitate max. 0.005 %
Chloride (Cl) max. 0.001 %
Iron (Fe) max. 0.001 %
Heavy Metals (as Pb) max. 0.001 %
Insoluble Matter and NH4OH Precipitate max. 0.01 %
Phosphate (PO4) max. 0.001 %
Trace Impurities (in ppm)
Arsenic (As) max. 1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR