Bis(1-butylpentyl) adipate ≥98,0%, Selectophore™ Ionophor, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

02150-25ML 02150-5ML
02150-25MLEA 424 EUR
02150-25ML 02150-5ML
Bis(1-butylpentyl) adipate ≥98,0%, Selectophore™ Ionophor, Supelco®
Bis(1-butylpentyl) adipate

Plasticizer employed in polymeric membrane electrodes.

Formel: C₂₄H₄₆O₄
Molecular Weight: 398.62 g/mol
Flammpunkt: 110 °C
MDL: MFCD00042947
CAS-Nummer: 77916-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4d9bf9a373dd29bcc95ed5975fc570049719c14&vid=f2d59b974bcd27ad228f5f2155b4f7cc10559d79&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR