Bis(1-butylpentyl) adipate ≥98,0%, Selectophore™ Ionophor, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
02150-25ML
02150-5ML
02150-25MLEA
424
EUR
02150-25ML
02150-5ML
Bis(1-butylpentyl) adipate ≥98,0%, Selectophore™ Ionophor, Supelco®
Bis(1-butylpentyl) adipate
Plasticizer employed in polymeric membrane electrodes.
Formel:
C₂₄H₄₆O₄ Molecular Weight: 398.62 g/mol Flammpunkt: 110 °C |
MDL:
MFCD00042947 CAS-Nummer: 77916-77-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...