2′,3′-Dideoxycytidine 5′-triphosphate sodium salt ≥98,0% (durch HPLC) Matrixsubstanz für MALDI-MS, Supelco®

Lieferant: Merck

Synonyme: ddCTP

M-Clarity™ Qualitätsstufe = MQ100

02241-1UMO-F
02241-1UMO-FEA 146 EUR
02241-1UMO-F
2′,3′-Dideoxycytidine 5′-triphosphate sodium salt ≥98,0% (durch HPLC) Matrixsubstanz für MALDI-MS, Supelco®
2′,3′-Dideoxycytidine 5′-triphosphate sodium salt

Inhibitor of DNA polymerase I-catalyzed chain elongation.

Formel: C₉H₁₆N₃O₁₂P₃
Molecular Weight: 451,16 g/mol
MDL: MFCD00717536
CAS-Nummer: 132619-66-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10b54f459a7cebf06032cf6efd5d9b180dd5642d&vid=d9c4cf8cbf134e3b08eec27c1851b408c15ad458&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Colorless
Appearance (Form) Liquid
Purity (HPLC area %) ≥ 98.0 %
HPLC minor component (area %) 2',3'-DIDEOXYCYTIDINE 5'-DIPHOSPHATE ≤1.5 %
Test by UV Spectroscopy 10.0 - 11.0 MMOL/L
Extraneous activities DNASES, RNASES NOT DETECTABLE
A (260NM) /A (280NM) Report Result
RECOMMENDED RETEST PERIOD 28 MONTHS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR