Oxalsäure Dihydrat 99,5-102,5%, Kristalle, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Ethandisäure Dihydrat

0230-07EA 0 EUR
0230-07
Oxalsäure Dihydrat 99,5-102,5%, Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Oxalsäure Dihydrat

Crystals. Lot analysis on label.


  • Lot analysis on label

Formel: HO₂CCO₂H·2H₂O
Molecular Weight: 126,07 g/mol
Siedepunkt: 149…160 °C (1013 hPa)
Schmelzpunkt: 101 °C
Dichte: 1,65 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149102
CAS-Nummer: 6153-56-6
EINECS: 205-634-3
UN: 3261
ADR: 8,III
REACH: 01-2119534576-33
Merck Index: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 99.5 - 102.5%
For Laboratory,Research,or Manufacturing Use ≤0.005%
Assay (HOCOCOOH2H₂O) (by KMnO₄ titrn) 99.5 - 102.5 %
Insoluble Matter ≤ 0.005 %
Residue after Ignition ≤ 0.010 %
Chloride (Cl) ≤ 0.002 %
Sulfate (SO₄) ≤ 0.002 %
Calcium (Ca)(by FES) ≤ 0.001 %
Nitrogen Compounds (as N) ≤ 0.001 %
Substances Darkened by Hot H₂SO₄ Passes Test
Trace Impurities - ACS - Heavy Metals (as Pb) ≤ 5 ppm
Trace Impurities - Iron (Fe) ≤ 2.0 ppm
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR