Natriumacetat Trihydrat, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
0256.0500EA
37.91
EUR
0256.0500
0256.1000
0256.5000
Natriumacetat Trihydrat, BAKER ANALYZED® ACS, J.T.Baker®
Natriumacetat Trihydrat
Formel:
H₃CCOONa·3H₂O Siedepunkt: 120…123 °C (1013 hPa) Schmelzpunkt: 58 °C Dichte: 1,4 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071557 CAS-Nummer: 6131-90-4 EINECS: 204-823-8 Merck Index: 13,08642 |
|
Spezifikation Testergebnisse
Meets ACS Specifications | |
Assay | 99.0 - 101.0 % |
Calcium (Ca) | max. 0.005 % |
Chloride (Cl) | max. 0.001 % |
Potassium (K) | max. 0.005 % |
Magnesium (Mg) | max. 0.002 % |
Insoluble Matter | max. 0.005 % |
pH of 5% Solution at 25°C | 7.5 - 9.2 |
Substances Reducing KMnO4 | passes test |
Sulfate (SO4) | max. 0.002 % |
Trace Impurities (in ppm) | |
Iron (Fe) | max. 5 |
Heavy Metals (as Pb) | max. 5 |
Phosphate (PO4) | max. 5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...