2',7'-Dichlorfluorescein -Lösung Indikator, anwendungsbereit, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

02591-500ML
02591-500MLEA 199.92 EUR
02591-500ML
2',7'-Dichlorfluorescein -Lösung Indikator, anwendungsbereit, Fluka™
2',7'-Dichlorfluorescein
Formel: C₂₀H₁₀Cl₂O₅
Molecular Weight: 401.20 g/mol
Siedepunkt: 81…83 °C
Schmelzpunkt: –90 °C (Hauptbestandteil)
Dichte: 0.79 g/cm³ (20 °C)
Flammpunkt: 12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005047
CAS-Nummer: 76-54-0
EINECS: 200-968-6
UN: 1219
ADR: 3 II

Bestellen

Spezifikation Testergebnisse

Appearance of the substance complying
Density (D 20/4) 0.775 - 0.804
Absorption maximum 505 - 519 nm
Absorbtion (1%/1cm) Max. 5.0
non-volatile matter Max. 0.001 %
water (Karl Fischer) Max. 0.1 %
titrable acid Max. 0.001 meq/g
aluminium (Al) Max. 0.5 ppm
boron (B) Max. 0.02 ppm
barium (Ba) Max. 0.1 ppm
bismuth (Bi) Max. 0.1 ppm
calcium (Ca) Max. 0.5 ppm
cadmium (Cd) Max. 0.05 ppm
cobalt (Co) Max. 0.02 ppm
chromium (Cr) Max. 0.02 ppm
copper (Cu) Max. 0.02 ppm
iron (Fe) Max. 0.1 ppm
potassium (K) Max. 0.5 ppm
lithium (Li) Max. 0.1 ppm
magnesium (Mg) Max. 0.1 ppm
manganese (Mn) Max. 0.02 ppm
molybdenum (Mo) Max. 0.1 ppm
sodium (Na) Max. 0.5 ppm
nickel (Ni) Max. 0.02 ppm
lead (Pb) Max. 0.1 ppm
tin (Sn) Max. 0.1 ppm
strontium (Sr) Max. 0.1 ppm
zinc (Zn) Max. 0.1 ppm
carbonyl compounds (as HCHO) Max. 100 ppm
APHA Max. 20


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR