di-Natriumtetraborat Decahydrat, BAKER, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate

0270.1000EA 53.16 EUR
0270.1000
di-Natriumtetraborat Decahydrat, BAKER, J.T.Baker®
di-Natriumtetraborat Decahydrat
Formel: Na₂B₄O₇·10H₂O
Molecular Weight: 381,37 g/mol
Siedepunkt: 320 °C (1013 hPa)
Schmelzpunkt: 75 °C
Dichte: 1,73 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149193
CAS-Nummer: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 13,08662

Bestellen

Spezifikation Testergebnisse

Meets Reagent Specifications for testing Ph. Eur. monographs.
Assay 99.0 - 103.0 %
Ammonium (NH4) max. 10 ppm
Appearance of solution passes test
Arsenic (As) max. 5 ppm
Calcium (Ca) max. 100 ppm
Heavy Metals (as Pb) max. 25 ppm
Identification passes test
pH 9.0 - 9.6
Sulfates (as SO4) max. 50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR