Sulfaethoxypyridazin, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

02743-50MG
02743-50MGEA 378 EUR
02743-50MG
Sulfaethoxypyridazin, Supelco®
Sulfaethoxypyridazin
Formel: C₁₂H₁₄N₄O₃S
MDL: MFCD00869228
CAS-Nummer: 963-14-4
EINECS: 213-514-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02bdccd1063baeba3caa914d256a0f40b89a36ba&vid=39b5c482b1613e134391d3acf270f5a18ae3d3cf&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR