tri-Natriumcitrat Dihydrat, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor

0280.1000EA 123.59 EUR
0280.1000 0280.0500 0280.5000
tri-Natriumcitrat Dihydrat, BAKER ANALYZED®, Laborreagens, J.T.Baker®
tri-Natriumcitrat Dihydrat
Formel: Na₃C₆H₅O₇·2H₂O
Schmelzpunkt: 150 °C
Dichte: 1,76 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150031
CAS-Nummer: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay min. 99.0 %
Calcium (Ca) max. 0.005 %
Chloride (Cl) max. 0.001 %
Insoluble Matter max. 0.005 %
pH of 5% Solution at 25°C 7.0 - 9.0
Sulfate (SO4) max. 0.002 %
Trace Impurities (in ppm) Iron (Fe) max. 5
Trace Impurities (in ppm) Heavy Metals (as Pb) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR