Pyrogallol, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Pyrogallussäure, 1,2,3-Trihydroxybenzol, 1,2,3-Benzoltriol

0289-04EA 0 EUR
0289-04
Pyrogallol, BAKER ANALYZED® ACS, J.T.Baker®
Pyrogallol
Formel: C₆H₆O₃
Molecular Weight: 126,11 g/mol
Siedepunkt: 309 °C (1013 hPa)
Schmelzpunkt: 130…135 °C
Dichte: 1,453 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002192
CAS-Nummer: 87-66-1
EINECS: 201-762-9
UN: 2811
ADR: 6.1,III
Merck Index: 13,08090
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09c5271be2848ebfa6e3614bff56b329a59cb6fe&vid=db25c87d92a56d9ff5d36f04d6bbf59acf236659&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Melting Point 131.0 - 135.0 °C
Residue after Ignition ≤0.005 %
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR