Blei (II)-oxid, körnig gold-, silberfrei

Lieferant: VWR Collection
Gefahr

029-930-11EA 0 EUR
029-930-11
Blei (II)-oxid, körnig gold-, silberfrei
Blei (II)-oxid

Litharge is a unique type of lead oxide used in the Fire Assay analysis of precious metals. It is produced from highly refined lead and offers a very low level of precious metal impurities.

Formel: PbO
Siedepunkt: 1470 °C (1013 hPa)
Schmelzpunkt: 888 °C
Dichte: 9,5 g/cm³ (20 °C)
MDL: MFCD00011164
CAS-Nummer: 1317-36-8
EINECS: 215-267-0
UN: 2291
ADR: 6.1,III
Merck Index: 13,05431

Bestellen

Spezifikation Testergebnisse

Appearance granularity Granular
Appearance colour Yellow to orange
Gold (Au) ∼ 1 ppb
Silver (Ag) max 0.16 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR