Natriumiodid, BAKER, Laboratory Reagent, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Iodnatrium

0294.0500EA 200.69 EUR
0294.0500
Natriumiodid, BAKER, Laboratory Reagent, J.T.Baker®
Natriumiodid
Formel: NaI
Molecular Weight: 149.89 g/mol
Siedepunkt: 1300
Schmelzpunkt: 662
Dichte: 3.67 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003532
CAS-Nummer: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9 III
Merck Index: 13,08705

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 100.5 %
Alkalinity passes test
Appearance of solution passes test
Iron (Fe) max. 20 ppm
Potassium (K) passes test
Heavy Metals (as Pb) max. 10 ppm
Identification passes test
Iodate (IO3) passes test
Limit of nitrate, nitrite and ammonia passes test
Loss on Drying max. 3.0 %
Organic Volatile Impurities passes test
Sulfate (SO4) max. 150 ppm
Thiosulfate and Barium passes test
Thiosulfates (as S2O3) passes test
Water (H2O) max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR