Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3), weiß ≥97%, Pulver

Lieferant: MP Biomedicals
Achtung

02EFC05591
02EFC05591EA 0 EUR
02EFC05591
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3), weiß ≥97%, Pulver
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3)

UTP has been used to study its vasodilatory effects on the skeletal muscle cells of diabetic patients[3]. UTP has also been used as a purinergic agonist to study ion transport functions of P2Y2 receptors on rat basolateral membranes.

Formel: C₉H₁₂N₂Na₃O₁₅P₃
MDL: MFCD00044310
CAS-Nummer: 19817-92-6
ADR: H302-H371,

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR