Stearinsäure triple pressed 90,0, powder NF, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
Synonyme:
Octadecansäure
0340-01EA
65.3
EUR
0340-01
Stearinsäure triple pressed 90,0, powder NF, J.T.Baker®
Stearinsäure
Triple pressed to remove impurities.
Formel:
C₁₈H₃₆O₂ Molecular Weight: 284.48 g/mol Siedepunkt: 361 °C (1013 hPa) Schmelzpunkt: 53…59 °C (als Gefrierpkt.) Dichte: 0.94 g/cm³ (20 °C) Flammpunkt: 196 °C (geschlossener Tiegel) Lagertemperatur: Kühlschrank |
MDL:
MFCD00002752 CAS-Nummer: 57-11-4 EINECS: 200-313-4 REACH: 01-2119537204-47 Merck Index: 13,08882 |
|
Spezifikation Testergebnisse
GMP Manufactured Product | Passes Test |
Meets N.F. Requirements | 194 - 212 |
CAUTION: For Manufacturing, processing or repackaging | Passes Test |
Bulk Pharmaceutical Chemical | ≤0.1% |
NF - Identification A | Passes Test |
NF - Identification B (Acid Value) | 194 - 212 |
NF - Identification C | Passes Test |
NF - Residue on Ignition | ≤0.1 % |
NF - Acidity | Passes Test |
NF - Color of Solution | Passes Test |
NF - Iodine Value | ≤4.0 |
NF - Freezing Point | 53 - 59 °C |
NF - Assay (C₁₈H₃₆O₂) (stearic acid) | 40.0 - 60.0 % |
NF - Assay (Total of Stearic & Palmitic) | ≥90.0 % |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...