Citronensäure 50% in wässriger Lösung, Biotechreagenz, J.T.Baker®

Lieferant: Avantor

0341-07EA 884.25 EUR
0341-07 0341-26 0341-27
Citronensäure 50% in wässriger Lösung, Biotechreagenz, J.T.Baker®
Citronensäure

Made from purified water USP and citric acid, anhydrous USP/ACS.

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 48 - 52%
CAUTION: For Manufacturing, processing or repackaging Passes Test
Assay (HOC(COOH)(CH₂COOH)₂) 48 - 52 %
Identification Passes Test
Chloride (Cl) ≤0.001 %
Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤3 ppm
Phosphate (PO₄) ≤0.001 %
Made from Purified Water U.S.P. and Citric Acid, anhydrous U.S.P./A.C.S.
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR