di-Natriumhydrogenphosphat Dihydrat, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor

0343.1000EA 95.41 EUR
0343.1000
di-Natriumhydrogenphosphat Dihydrat, BAKER ANALYZED®, Laborreagens, J.T.Baker®
di-Natriumhydrogenphosphat Dihydrat
Formel: Na₂HPO₄·2H₂O
MDL: MFCD00149182
CAS-Nummer: 10028-24-7

Bestellen

Spezifikation Testergebnisse

Assay min. 99 %
Chloride (Cl) max. 0.001 %
Iron (Fe) max. 0.001 %
Heavy Metals (as Pb) max. 0.001 %
Insoluble Matter max. 0.005 %
pH of 0.1 M Solution at 25°C 9.0 - 9.5
Sulfate (SO4) max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR