Salzsäure 6.0 N, Biotechreagenz, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Hydrochloric acid solution, Muriatic acid
Veränderungsmanagement-Kategorie= R
0347-02EA
0
EUR
0347-02
Salzsäure 6.0 N, Biotechreagenz, J.T.Baker®
Salzsäure
Formel:
HCl Molecular Weight: 36,46 g/mol Siedepunkt: 2230 °C (1013 hPa) Schmelzpunkt: 1710 °C Dichte: 1,06 g/cm³ |
MDL:
MFCD00011324 CAS-Nummer: 7647-01-0 UN: 1789 ADR: 8,II |
Spezifikation Testergebnisse
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Appearance (Clear, colorless liquid) | Passes Test |
Normality | 5.95 - 6.05 |
Ammonium (NH₄) | ≤ 3 ppm |
Trace Impurities - Heavy Metals | ≤ 0.5 ppm |
Trace Impurities - Iron (Fe) | ≤ 0.1 ppm |
Sulfate (SO₄) | ≤ 1 ppm |
Made from U.S.P. Water for Injection (WFI) and Hydrochloric Acid, N.F. | |
(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical | |
Specifications | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Filtered through a 0.2 micron filter. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...