Sulfanilsäure, wasserfrei 98,0-102,0%, Pulver, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

0354-01EA 0 EUR
0354-01
Sulfanilsäure, wasserfrei 98,0-102,0%, Pulver, BAKER ANALYZED® ACS, J.T.Baker®
Sulfanilsäure

For nitrogen dioxide determination (ASTM D-1607).


  • Lot analysis on label

Formel: 4-(H₂N)C₆H₄SO₃H
Siedepunkt: 363 °C (1013 hPa)
Schmelzpunkt: 279 °C
Dichte: 1,485 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007886
CAS-Nummer: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 98.0 - 102.0%
For Laboratory,Research,or Manufacturing Use ≤0.010%
Assay (4-NH₂C₆H₄SO₃H) 98.0 - 102.0 %
Residue after Ignition ≤ 0.010 %
Insoluble in Na₂CO₃ Solution ≤ 0.01 %
Chloride (Cl) ≤ 0.002 %
Sulfate (SO₄) ≤ 0.005 %
Suitability for Nitrogen Dioxide Determination (ASTM D-1607) Passes Test
Nitrite (NO₂) ≤ 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR