5-Sulfosalicylsäure Dihydrat 99,0-101,0%, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

0364-05EA 1545.16 EUR
0364-05 0364-01
5-Sulfosalicylsäure Dihydrat 99,0-101,0%, BAKER ANALYZED® ACS, J.T.Baker®
5-Sulfosalicylsäure Dihydrat

  • Lot analysis on label

Formel: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
Schmelzpunkt: 120 °C
Dichte: 1,695 g/cm³ (20 °C)
Flammpunkt: 150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149540
CAS-Nummer: 5965-83-3
EINECS: 202-555-6
UN: 2585
ADR: 8,III
Merck Index: 13,09054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69e00688850e885438e2fdd9a62f6d564ebf6ae8&vid=6a3840cf38254959a0062041e9a84fa6740b1288&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 99.0 - 101.0%
For Laboratory,Research,or Manufacturing Use ≤0.02%
Assay (C₇H₆O₆S2H₂O) 99.0 - 101.0 %
Insoluble Matter ≤ 0.02 %
Residue after Ignition ≤ 0.10 %
Chloride (Cl) ≤ 0.001 %
Salicylic Acid (HOC₆H₄COOH) ≤ 0.04 %
Sulfate (SO₄) ≤ 0.02 %
Heavy Metals (as Pb) ≤ 0.002 %
Trace Impurities - Iron (Fe) ≤ 0.001 %
STORAGE: Keep product from light.
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR