Nitrite ionophore VI, Selectophore™, Funktion getestet Ionophor, Supelco®

Lieferant: Merck

03721-250MG
03721-250MGEA 849 EUR
03721-250MG
Nitrite ionophore VI, Selectophore™, Funktion getestet Ionophor, Supelco®
Nitrite ionophore VI
Formel: C₃₆H₄₆CoN₂O₂
CAS-Nummer: 228577-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR