Organic reference standard, gamma-Aminobutyric acid, analytical standard, Supelco®
Lieferant: Merck
03835-250MG
03835-250MGEA
61.6
EUR
03835-250MG
Organic reference standard, gamma-Aminobutyric acid, analytical standard, Supelco®
Standards
Organische Standards
γ-Aminobutyric acid is a chief inhibitory neurotransmitter found in the cerebellum, hypothalamus, thalamus and hippocampus. It is formed via the decarboxylation of L-glutamate catalyzed by the enzyme, glutamic acid decarboxylase (GAD).
The major inhibitory neurotransmitter in brain are GABAA and GABAB receptor agonist and increased Cl−
conductance.