Organic reference standard, gamma-Aminobutyric acid, analytical standard, Supelco®
Lieferant: Merck
03835-250MG
03835-250MGEA 61.6 EUR
03835-250MG
Organic reference standard, gamma-Aminobutyric acid, analytical standard, Supelco®
Standards Organische Standards
γ-Aminobutyric acid is a chief inhibitory neurotransmitter found in the cerebellum, hypothalamus, thalamus and hippocampus. It is formed via the decarboxylation of L-glutamate catalyzed by the enzyme, glutamic acid decarboxylase (GAD).
The major inhibitory neurotransmitter in brain are GABAA and GABAB receptor agonist and increased Cl−
conductance.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cd105ebc5810fd51d65149c97d5c22f31d01205&vid=76d92f035934ffeffff1611db45ee59045606cdf&prid=web&clid=SAPDAM