Natriumhydroxid 2.0 N in wässriger Lösung, Biotechreagenz, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Caustic soda

Filtriert über 0,2 µm-Filter

0390-07EA 1509.26 EUR
0390-07
Natriumhydroxid 2.0 N in wässriger Lösung, Biotechreagenz, J.T.Baker®
Sodium hydroxide

Made from water for injection USP and sodium hydroxide pellets, NF (multi-compendial) which meets BP, Ph. Eur. and JP chemical specifications.

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 109 °C (1013 hPa)
Dichte: ≤1,11 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 1.9 - 2.1
Protect from air to avoid absorption of carbon dioxide. ≤5 ppm
CAUTION: For Manufacturing, processing or repackaging ≤1 ppm
Appearance (Clear, colorless liquid) Passes Test
Normality 1.9 - 2.1
Chloride (Cl) ≤ 5 ppm
ACS - Heavy Metals (as Pb) ≤ 1 ppm
Trace Impurities - Iron (Fe) ≤ 0.5 ppm
Endotoxin Concentration (EU/mL ≤ 2
Made from Water for Injection U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR