Kaliumchlorid 99-100,5% ACS
Lieferant: VWR Chemicals
Synonyme:
Potassiumchloride
0395-1KGEA
166
EUR
0395-1KG
0395-2.5KG
0395-500G
0395-12KG
Kaliumchlorid 99-100,5% ACS
Potassium chloride
Formel:
KCl Molecular Weight: 74.55 g/mol Siedepunkt: 1420 Schmelzpunkt: 773 Dichte: 1.98 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011360 CAS-Nummer: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Spezifikation Testergebnisse
Barium Test | Pass |
Bromide | ≤ 0,01% |
Calcium | ≤ 0,002% |
Chlorate & Nitrate (as NO3) | ≤ 0,003% |
Heavy Metals | ≤ 0,0005% |
Insolubles | ≤ 0,005% |
Iodide | ≤ 0,002% |
Iron | ≤ 0,0003% |
Magnesium | ≤ 0,001% |
pH (5%, Wasser)@25°C | 5,4 - 8,6 |
Phosphate | ≤ 0,0005% |
Purity | 99 - 100,5% |
Sodium | ≤ 0,005% |
Sulphate | ≤ 0,001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...