Kaliumchlorid 99-100,5% ACS

Lieferant: VWR Chemicals

Synonyme: Potassiumchloride

0395-1KGEA 166 EUR
0395-1KG 0395-2.5KG 0395-500G 0395-12KG
Kaliumchlorid 99-100,5% ACS
Potassium chloride
Formel: KCl
Molecular Weight: 74.55 g/mol
Siedepunkt: 1420
Schmelzpunkt: 773
Dichte: 1.98 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011360
CAS-Nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Bestellen

Spezifikation Testergebnisse

Barium Test Pass
Bromide ≤ 0,01%
Calcium ≤ 0,002%
Chlorate & Nitrate (as NO3) ≤ 0,003%
Heavy Metals ≤ 0,0005%
Insolubles ≤ 0,005%
Iodide ≤ 0,002%
Iron ≤ 0,0003%
Magnesium ≤ 0,001%
pH (5%, Wasser)@25°C 5,4 - 8,6
Phosphate ≤ 0,0005%
Purity 99 - 100,5%
Sodium ≤ 0,005%
Sulphate ≤ 0,001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR