Boc-Trp(Boc)-OH ≥98% (durch TLC, NMR), Novabiochem®

Lieferant: NOVABIOCHEM

Synonyme: N-Boc-1-Boc-L-tryptophan, (S)-3-(1-(tert-Butoxycarbonyl)-1H-indol-3-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid

8.53078.0025
04-12-0205-25EA 641 EUR
04-12-0205-25
Boc-Trp(Boc)-OH ≥98% (durch TLC, NMR), Novabiochem®
Boc-Trp(Boc)-OH
Formel: C₂₁H₂₈N₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00270520
CAS-Nummer: 144599-95-1

Bestellen

Spezifikation Testergebnisse

Colour (Visual) White to Slight Yellow to Beige
Appearance of Substance (Visual) Powder
Identity (IR) Passes Test
Optical Rotation Α 25/D (C=1 in Methanol) +13.5 - +17.5°
Purity (TLC(011A)) ≥98%
Purity (TLC(0811)) ≥98%
Solubility (1 mmole in 2 ml DMF) Clear Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR