Boc-L-Arg(Z)2-OH ≥98,0% (durch HPLC), Novabiochem®

Lieferant: NOVABIOCHEM

Synonyme: Boc-Arg(Cbz)₂-OH, Boc-Arg(Z)2-OH

8.53036.0005
04-12-0220-5EA 199 EUR
04-12-0220-5
Boc-L-Arg(Z)2-OH ≥98,0% (durch HPLC), Novabiochem®
Boc-L-Arg(Z)2-OH
Formel: C₂₇H₃₄N₄O₈
Molecular Weight: 542,6 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00236824
CAS-Nummer: 51219-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR