Fmoc-Met-OH
Lieferant: NOVABIOCHEM
Synonyme:
N-Fmoc-L-methionine, FMOC-L-Methionine
8.52002.0100
04-12-1003-100EA
68.2
EUR
04-12-1003-100
Fmoc-Met-OH
Fmoc-Met-OH
Formel:
C₂₀H₂₁NO₄S Molecular Weight: 371.46 g/mol |
MDL:
MFCD00037134 CAS-Nummer: 71989-28-1 EINECS: 276-258-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...