N-Fmoc-β-alanin

Lieferant: NOVABIOCHEM

Synonyme: Fmoc-β-Ala-OH P, Fmoc-β-Ala-p-alkoxybenzyl alcohol, N-Fmoc-beta-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-beta-alanine

8.52024.0025 8.52024.0005
04-12-1044-25EA 195 EUR
04-12-1044-25 04-12-1044-5
N-Fmoc-β-alanin
N-Fmoc-β-alanin
Formel: C₁₈H₁₇NO₄
Molecular Weight: 311.3 g/mol
MDL: MFCD00063328
CAS-Nummer: 35737-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR