Fmoc-Thi-OH

Lieferant: NOVABIOCHEM

8.52039.0001 8.52039.0005
04-12-1082-1EA 150 EUR
04-12-1082-1 04-12-1082-5
Fmoc-Thi-OH
Fmoc-Thi-OH
Formel: C₂₂H₁₉NO₄S
MDL: MFCD00065676
CAS-Nummer: 130309-35-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR