Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl)

Lieferant: NOVABIOCHEM

Synonyme: Methyl glycinate hydrochloride, H-Gly-OMe.HCl, Glycine Methyl Ester HCl

8.54066.0025
04-12-5007-25EA 0 EUR
04-12-5007-25
Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl)
Glycinmethylester Hydrochlorid (H-Gly-OMe.HCl)
Formel: C₃H₈ClNO₂
Molecular Weight: 125,56 g/mol
MDL: MFCD00012870
CAS-Nummer: 5680-79-5
EINECS: 227-139-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR