N-Boc-D-valin
Lieferant: NOVABIOCHEM
Synonyme:
Boc-D-Val-OH
8.53098.0025
04-13-0031-25EA
182
EUR
04-13-0031-25
N-Boc-D-valin
N-Boc-D-valin
Formel:
C₁₀H₁₉NO₄ Molecular Weight: 217,3 g/mol |
MDL:
MFCD00038282 CAS-Nummer: 22838-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...