N(α)-Fmoc-D-tryptophan

Lieferant: NOVABIOCHEM

8.52150.0025 8.52150.0005
04-13-1035-25EA 566 EUR
04-13-1035-25 04-13-1035-5
N(α)-Fmoc-D-tryptophan
N(α)-Fmoc-D-tryptophan
Formel: C₂₆H₂₂N₂O₄
MDL: MFCD00062954
CAS-Nummer: 86123-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR