Natriumhydroxid, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Caustic soda

0403.5000EA 193.42 EUR
0403.5000
Natriumhydroxid, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Sodium hydroxide
Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1390 °C (1013 hPa)
Schmelzpunkt: 323 °C
Dichte: 2,13 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Bestellen

Spezifikation Testergebnisse

Assay min. 99.0 %
Ammonium Hydroxide Precipitate max. 0.02 %
Chloride (Cl) max. 5 ppm
Potassium (K) max. 0.01 %
Heavy Metals (as Ag) max. 5 ppm
Heavy Metals (as Pb) max. 5 ppm
Na2CO3 max. 1.0 %
Trace Impurities (in ppm)
Iron (Fe) max. 3
Nickel (Ni) max. 5
Mercury (Hg) (by AAS) max. 0.1
Nitrogen Compounds (as N) max. 3
Phosphate (as PO4) max. 10
Sulfate (SO4) max. 5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR