Melting point standards, sodium methanesulfonate, traceable to primary standards, Supelco®
Lieferant: Merck
04229-5G
04229-5GEA
50.1
EUR
04229-5G
Melting point standards, sodium methanesulfonate, traceable to primary standards, Supelco®
Standards
Schmelzpunkt-Standards
Mettler-Toledo Calibration substance ME 30130610, Sodium methanesulfonate, traceable to primary standards (LGC).
Suitable for dropping point calibration.