Melting point standards, sodium methanesulfonate, traceable to primary standards, Supelco®
Lieferant: Merck
04229-5G
04229-5GEA 50.1 EUR
04229-5G
Melting point standards, sodium methanesulfonate, traceable to primary standards, Supelco®
Standards Schmelzpunkt-Standards
Mettler-Toledo Calibration substance ME 30130610, Sodium methanesulfonate, traceable to primary standards (LGC).
Suitable for dropping point calibration.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4254147b0ed9ca24e27c9a7eebcd0a8b1880e00&vid=d4254147b0ed9ca24e27c9a7eebcd0a8b1880e00&prid=web&clid=SAPDAM