Bromthymolblau ACS

Lieferant: VWR Chemicals

0431-25GEA 200 EUR
0431-25G
Bromthymolblau ACS
Bromthymolblau

pH indicator goes from yellow to blue over a range of 6.0 to 7.6. Used to trace movement of fluids from the lymph and also for demonstration of fungal hyphae within plant roots.

Formel: C₂₇H₂₈Br₂O₅S
Schmelzpunkt: 204…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005872
CAS-Nummer: 76-59-5
EINECS: 200-971-2
Merck Index: 13,01430

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR