Organic reference standard, Bisphenol E, analytical standard, Supelco®
Lieferant: Merck
04487-100MG
04487-100MGEA
52.8
EUR
04487-100MG
Organic reference standard, Bisphenol E, analytical standard, Supelco®
Standards
Organische Standards
Bisphenol E may be used as a reference standard in the determination of bisphenol E in beverages using on-line solid-phase extraction with fast liquid chromatography coupled with tandem mass spectrometry (on-line SPE fast LC-MS/MS).