Supelco® Organic Reference Standard, Digoxin
Lieferant: Merck
04599-50MG
04599-50MGEA
86.4
EUR
04599-50MG
Supelco® Organic Reference Standard, Digoxin
Standards
Organische Standards
- GHS signal: GHS06
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Bitte beachten P260 - P264 - P270 - P301 + P310 - P304 + P340 + P311 - P314