1,10-Phenanthrolin Monohydrat ACS

Lieferant: VWR Chemicals
Gefahr

Synonyme: 1,10-Phenanthroline hydrate

0516-10GEA 106 EUR
0516-10G
1,10-Phenanthrolin Monohydrat ACS
1,10-Phenanthrolin Monohydrat

A metalloprotease inhibitor useful for chelating iron and other divalent metals.

Formel: C₁₂H₈N₂·1H₂O
Molecular Weight: 198,22 g/mol
Schmelzpunkt: 93…94 °C
Dichte: 1,259 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149973
CAS-Nummer: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Suitability as Redox Indicator Pass
Suitability for Determining Iron pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR