Erythromycin A enol ether, analytical standard, Supelco®
Lieferant: Merck
05238-5MG
05238-5MGEA
583
EUR
05238-5MG
Erythromycin A enol ether, analytical standard, Supelco®
Standards
Organische Standards
Erythromycin A enol ether (EAEN) is an acidic degradation product of erythromycin A (EA) which is grouped under the macrolide antibiotic class of compounds.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.