Erythromycin A enol ether, analytical standard, Supelco®
Lieferant: Merck
05238-5MG
05238-5MGEA 583 EUR
05238-5MG
Erythromycin A enol ether, analytical standard, Supelco®
Standards Organische Standards
Erythromycin A enol ether (EAEN) is an acidic degradation product of erythromycin A (EA) which is grouped under the macrolide antibiotic class of compounds.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fda1c1847deb8e5f47b2ba4653c1906248b16db&vid=9fda1c1847deb8e5f47b2ba4653c1906248b16db&prid=web&clid=SAPDAM