Supelco® Organic Reference Standard, Acenaphthene
Lieferant: Merck
05426-100MG
05426-100MGEA 85 EUR
05426-100MG
Supelco® Organic Reference Standard, Acenaphthene
Standards Organische Standards
  • GHS signal: GHS07, GHS09
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P273 - P391 - P501
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acc11d3de0469c9f8fce9cdf28139ae6978b48f2&vid=acc11d3de0469c9f8fce9cdf28139ae6978b48f2&prid=web&clid=SAPDAM